Assay Details
| Gene Symbol: Adrbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290818.1 | NP_001277747.1 | 19 - 20 | 2001 | 114 | |
| NM_130863.2 | NP_570933.1 | 20 - 21 | 2073 | 114 | ||
| GenBank mRNA | AK155691.1 | - | 19 - 20 | 2002 | 114 | |
| BC003196.1 | - | 19 - 20 | 1789 | 114 | ||
| BC033272.1 | - | 16 - 17 | 2149 | 114 | ||
| BC038312.1 | - | 1 - 2 | 672 | 114 | ||
| BC053922.1 | - | 20 - 21 | 1806 | 6309555 | 114 | |
More Information
Set Membership: |
|

Adrbk1
Genomic Map