Assay Details
| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134009.3 | NP_598770.1 | 11 - 12 | 1597 | 89 | |
| GenBank mRNA | AK048954.1 | - | 11 - 12 | 1591 | 89 | |
| BC019501.1 | - | 11 - 12 | 1597 | 4193809 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncln
Genomic Map