Assay Details
| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134009.3 | NP_598770.1 | 12 - 13 | 1648 | 86 | |
| GenBank mRNA | AK019409.1 | - | 12 - 13 | 1517 | 85 | |
| AK048954.1 | - | 12 - 13 | 1642 | 86 | ||
| AK181997.1 | - | 1 - 2 | 51 | 86 | ||
| AK209854.1 | - | 1 - 2 | 51 | 86 | ||
| BC019501.1 | - | 12 - 13 | 1648 | 4193809 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncln
Genomic Map