Assay Details
| Gene Symbol: Ncln | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134009.3 | NP_598770.1 | 5 - 6 | 802 | 72 | |
| GenBank mRNA | AK019409.1 | - | 5 - 6 | 672 | 72 | |
| AK048954.1 | - | 5 - 6 | 796 | 72 | ||
| AK128887.1 | - | 5 - 6 | 803 | 72 | ||
| BC019501.1 | - | 5 - 6 | 802 | 4193809 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncln
Genomic Map