Assay Details
| Gene Symbol: Kdm4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161823.1 | NP_001155295.1 | 14 - 15 | 2439 | 94 | |
| NM_172382.2 | NP_759014.2 | 14 - 15 | 2439 | 94 | ||
| GenBank mRNA | AK054095.1 | - | 14 - 15 | 2403 | 94 | |
| AK129187.1 | - | 14 - 15 | 2294 | 94 | ||
| AK136085.1 | - | 5 - 6 | 1096 | 94 | ||
| AK145066.1 | - | 14 - 15 | 2425 | 94 | ||
| AK145947.1 | - | 14 - 15 | 2361 | 94 | ||
| BC028866.1 | - | 14 - 15 | 2342 | 4207552 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kdm4a
Genomic Map