Assay Details
| Gene Symbol: Nod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171007.1 | NP_001164478.1 | 7 - 8 | 2668 | 61 | |
| NM_172729.3 | NP_766317.1 | 7 - 8 | 2919 | 61 | ||
| GenBank mRNA | AK082663.1 | - | 4 - 5 | 1714 | 61 | |
| AK089662.1 | - | 7 - 8 | 2669 | 61 | ||
| AK137585.1 | - | 7 - 8 | 2924 | 61 | ||
| AY160222.1 | - | 6 - 7 | 2454 | 61 | ||
| BC042670.1 | - | 7 - 8 | 2746 | 3597911 | 61 | |
| BC043670.1 | - | 4 - 5 | 1636 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nod1
Genomic Map