Assay Details
| Gene Symbol: Lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175413.3 | NP_780622.1 | 7 - 8 | 1045 | 66 | |
| GenBank mRNA | AK039277.1 | - | 7 - 8 | 1042 | 66 | |
| AK052159.1 | - | 7 - 8 | 997 | 66 | ||
| AK079125.1 | - | 7 - 8 | 1023 | 66 | ||
| BC119529.2 | - | 7 - 8 | 994 | 40056120 | 66 | |
| BC119530.1 | - | 7 - 8 | 995 | 40056121 | 66 | |
More Information
Set Membership: |
|

Lrrc39
Genomic Map