Assay Details
| Gene Symbol: Adamts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277305.1 | NP_001264234.1 | 14 - 15 | 2285 | 107 | |
| NM_175643.3 | NP_783574.1 | 14 - 15 | 2285 | 107 | ||
| GenBank mRNA | AK040370.1 | - | 8 - 9 | 1029 | 107 | |
| AK220464.1 | - | 13 - 14 | 2027 | 107 | ||
| BC046456.1 | - | 14 - 15 | 2285 | 3965027 | 107 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts2
Genomic Map