Assay Details
| Gene Symbol: Adamts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277305.1 | NP_001264234.1 | 17 - 18 | 2695 | 62 | |
| NM_175643.3 | NP_783574.1 | 17 - 18 | 2695 | 62 | ||
| GenBank mRNA | AK040370.1 | - | 11 - 12 | 1439 | 62 | |
| AK220464.1 | - | 16 - 17 | 2437 | 62 | ||
| BC046456.1 | - | 17 - 18 | 2695 | 3965027 | 62 | |
| BC115840.1 | - | 4 - 5 | 438 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adamts2
Genomic Map