Assay Details
| Gene Symbol: Adamts2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277305.1 | NP_001264234.1 | 6 - 7 | 1209 | 74 | |
| NM_175643.3 | NP_783574.1 | 6 - 7 | 1209 | 74 | ||
| GenBank mRNA | AK220464.1 | - | 5 - 6 | 951 | 74 | |
| BC046456.1 | - | 6 - 7 | 1209 | 3965027 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Adamts2
Genomic Map