Assay Details
| Gene Symbol: Mysm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177239.2 | NP_796213.2 | 3 - 4 | 240 | 153 | |
| GenBank mRNA | AK043802.1 | - | 3 - 4 | 240 | 153 | |
| AK081684.1 | - | 3 - 4 | 215 | 153 | ||
| AK164103.1 | - | 3 - 4 | 186 | 153 | ||
| BC150946.1 | - | 3 - 4 | 236 | 9087857 | 153 | |
| BC151172.1 | - | 3 - 4 | 236 | 9088061 | 153 | |
More Information
Set Membership: |
|

Mysm1
Genomic Map