Assay Details
| Gene Symbol: Pik3r5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177320.2 | NP_796294.2 | 16 - 17 | 2432 | 69 | |
| GenBank mRNA | AK149847.1 | - | 16 - 17 | 2400 | 69 | |
| AK154643.1 | - | 16 - 17 | 2411 | 69 | ||
| AK161050.1 | - | 7 - 8 | 1340 | 69 | ||
| AK171279.1 | - | 15 - 16 | 2557 | 69 | ||
| AY156924.1 | - | 15 - 16 | 2281 | 69 | ||
| BC128076.1 | - | 15 - 16 | 2281 | 40060132 | 69 | |
| BC128077.1 | - | 15 - 16 | 2281 | 40060133 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pik3r5
Genomic Map