Assay Details
| Gene Symbol: Gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178406.2 | NP_848493.1 | 4 - 5 | 1118 | 76 | |
| GenBank mRNA | BC029098.1 | - | 3 - 4 | 1024 | 3481871 | 76 |
| BC037650.1 | - | 3 - 4 | 1117 | 5151632 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417688 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417642 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417716 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417646 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417712 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417641 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417711 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417645 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417710 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417643 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417687 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110998 | BC037650.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110998 | NM_178406.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110998 | AK004377.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110998 | BC019401.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 568608 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422544 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 336434 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478372 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 937257 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 625938 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714216 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 139296 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8108 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P153943 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110998 | BC037650.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110998 | NM_178406.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110998 | AK004377.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110998 | BC019401.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417661 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417657 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417664 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417646 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417655 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417667 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417697 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417658 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417640 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417654 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417651 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417693 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417642 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417666 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417662 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417648 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23417682 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P153943 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05454589 | - | - |
| Whole Mouse Genome G4122A | A_52_P48311 | BC037650.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48311 | BC029098.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48311 | NM_178406.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48311 | AK004377.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P48311 | BC019401.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105853 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P153943 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03303575 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03303591 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417642 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417712 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417711 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417710 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417688 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417641 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417690 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417716 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417643 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417689 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417715 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417687 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417646 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417713 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23417645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417704 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417731 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417794 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417743 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417727 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417765 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417682 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417799 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417797 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417697 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417779 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23417785 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417727 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417689 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417690 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03536373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417651 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03465927 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417723 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417680 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03634607 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417686 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417678 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417685 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03610235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417654 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03634616 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03303575 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417693 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417682 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417625 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417695 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03536358 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417681 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417691 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417638 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417667 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03634614 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417698 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03326909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03403594 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417643 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417646 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417672 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03326905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417664 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417731 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03536371 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23417729 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10510703 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510707 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510702 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510705 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510701 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510706 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10510704 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1426973_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10510700 | - | - |
| Mouse Genome 430 2.0 Array | 1426973_at | - | - |
| Mouse Genome 430 2.0 Array | 1426973_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100129_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 100129_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400016541.mm.1 | NM_178406.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016541.mm.1 | NM_178406.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400016541.mm.1 | TC0400002018.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1426973_PM_at | - | - |
| Mouse Expression Set 430 | 1426973_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426973_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1426973_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426973_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1426973_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 880437 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4120332 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4120332 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004120332 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2590975 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2590975 | - | - |
| mouse-6 v1.1 (Illumina) | scl24655.6_237-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4120332 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24655.6_237-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4120332 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2590975 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2590975 | - | - |

Gpr153
Genomic Map