Assay Details
| Gene Symbol: Hadha | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178878.2 | NP_849209.1 | 7 - 8 | 704 | 117 | |
| GenBank mRNA | AK029017.1 | - | 7 - 8 | 705 | 117 | |
| AK035399.1 | - | 7 - 8 | 687 | 117 | ||
| AK053986.1 | - | 7 - 8 | 702 | 117 | ||
| AK150188.1 | - | 7 - 8 | 702 | 117 | ||
| AK170478.1 | - | 7 - 8 | 705 | 117 | ||
| AK170683.1 | - | 7 - 8 | 705 | 117 | ||
| AK171469.1 | - | 7 - 8 | 705 | 117 | ||
| BC037009.1 | - | 7 - 8 | 692 | 5134605 | 117 | |
| BC046978.1 | - | 7 - 8 | 678 | 6315204 | 117 | |
| BC058569.1 | - | 7 - 8 | 700 | 6468514 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hadha
Genomic Map