| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500009109.mm.1 |
TC0500001079.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009109.mm.1 |
NM_175241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009109.mm.1 |
NM_175241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009109.mm.1 |
NM_182745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009109.mm.1 |
NM_182745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009112.mm.1 |
TC0500001079.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009112.mm.1 |
NM_175241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009112.mm.1 |
NM_175241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009112.mm.1 |
NM_182745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009112.mm.1 |
NM_182745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009110.mm.1 |
TC0500001079.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009110.mm.1 |
NM_175241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009110.mm.1 |
NM_175241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009110.mm.1 |
NM_182745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009110.mm.1 |
NM_182745.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009114.mm.1 |
TC0500001079.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500009114.mm.1 |
NM_175241.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009114.mm.1 |
NM_175241.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009114.mm.1 |
NM_182745.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500009114.mm.1 |
NM_182745.2 |
Refseq |