Assay Details
| Gene Symbol: Ssrp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136081.2 | NP_001129553.1 | 5 - 6 | 739 | 75 | |
| NM_182990.4 | NP_892035.2 | 5 - 6 | 868 | 75 | ||
| GenBank mRNA | AK146446.1 | - | 5 - 6 | 791 | 75 | |
| AK146585.1 | - | 5 - 6 | 788 | 75 | ||
| AK146607.1 | - | 5 - 6 | 771 | 75 | ||
| AK146691.1 | - | 5 - 6 | 771 | 75 | ||
| AK147005.1 | - | 5 - 6 | 801 | 75 | ||
| AK151584.1 | - | 5 - 6 | 821 | 75 | ||
| AK158552.1 | - | 5 - 6 | 739 | 75 | ||
| AK183865.1 | - | 2 - 3 | 75 | |||
| AK201898.1 | - | 2 - 3 | 75 | |||
| BC004848.1 | - | 5 - 6 | 735 | 75 | ||
| BC024835.1 | - | 2 - 3 | 238 | 75 | ||
| BC042502.1 | - | 5 - 6 | 716 | 5005209 | 75 | |
| S50213.1 | - | 4 - 5 | 613 | 75 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ssrp1
Genomic Map