Assay Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183019.2 | NP_898840.2 | 5 - 6 | 1053 | 66 | |
| GenBank mRNA | AK034013.1 | - | 5 - 6 | 1017 | 66 | |
| AK122448.2 | - | 1 - 2 | 108 | 66 | ||
| AK138598.1 | - | 5 - 6 | 1013 | 66 | ||
| AK163424.1 | - | 5 - 6 | 1035 | 66 | ||
| AK169957.1 | - | 5 - 6 | 1073 | 66 | ||
| BC055804.1 | - | 5 - 6 | 1014 | 66 | ||
| BC094608.1 | - | 4573 | 66 | |||
| BC139371.1 | - | 5 - 6 | 1036 | 8862393 | 66 | |
| BC139397.1 | - | 5 - 6 | 1036 | 8862419 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef4
Genomic Map