Assay Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183029.2 | NP_898850.2 | 10 - 11 | 1521 | 82 | |
| GenBank mRNA | AK044984.1 | - | 9 - 10 | 1037 | 82 | |
| AK049196.1 | - | 10 - 11 | 1217 | 82 | ||
| AK077404.1 | - | 10 - 11 | 1202 | 82 | ||
| AK159654.1 | - | 10 - 11 | 1254 | 82 | ||
| AK170531.1 | - | 9 - 10 | 1025 | 82 | ||
| AY531659.1 | - | 10 - 11 | 1185 | 82 | ||
| BC023758.1 | - | 10 - 11 | 1219 | 82 | ||
| BC054552.1 | - | 10 - 11 | 1241 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp2
Genomic Map