Assay Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183029.2 | NP_898850.2 | 12 - 13 | 1696 | 67 | |
| GenBank mRNA | AK044984.1 | - | 11 - 12 | 1212 | 67 | |
| AK049196.1 | - | 12 - 13 | 1392 | 67 | ||
| AK077404.1 | - | 12 - 13 | 1377 | 67 | ||
| AK159654.1 | - | 12 - 13 | 1429 | 67 | ||
| AK170531.1 | - | 11 - 12 | 1200 | 67 | ||
| AY531659.1 | - | 12 - 13 | 1360 | 67 | ||
| BC023758.1 | - | 12 - 13 | 1394 | 67 | ||
| BC054552.1 | - | 12 - 13 | 1416 | 67 | ||
| BU708344.1 | - | 2 - 3 | 141 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp2
Genomic Map