Assay Details
| Gene Symbol: Igf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183029.2 | NP_898850.2 | 2 - 3 | 575 | 93 | |
| GenBank mRNA | AK044984.1 | - | 1 - 2 | 91 | 93 | |
| AK049196.1 | - | 2 - 3 | 272 | 93 | ||
| AK077404.1 | - | 2 - 3 | 254 | 93 | ||
| AK159654.1 | - | 2 - 3 | 310 | 93 | ||
| AY531659.1 | - | 2 - 3 | 239 | 93 | ||
| BC023758.1 | - | 2 - 3 | 273 | 93 | ||
| BC054552.1 | - | 2 - 3 | 295 | 93 | ||
| CB290059.1 | - | 2 - 3 | 575 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Igf2bp2
Genomic Map