Assay Details
| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183089.2 | NP_898912.2 | 6 - 7 | 836 | 97 | |
| GenBank mRNA | AK008137.1 | - | 6 - 7 | 836 | 97 | |
| AK011162.1 | - | 6 - 7 | 853 | 97 | ||
| BC099382.1 | - | 5 - 6 | 465 | 97 | ||
| BC116833.1 | - | 6 - 7 | 811 | 40126152 | 97 | |
| BC116835.1 | - | 6 - 7 | 811 | 40126154 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dscc1
Genomic Map