Assay Details
| Gene Symbol: Dscc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183089.2 | NP_898912.2 | 8 - 9 | 1142 | 74 | |
| GenBank mRNA | AK008137.1 | - | 8 - 9 | 1142 | 74 | |
| AK011162.1 | - | 8 - 9 | 1159 | 74 | ||
| AK144502.1 | - | 9 - 10 | 1334 | 74 | ||
| BC099382.1 | - | 7 - 8 | 771 | 74 | ||
| BC116833.1 | - | 8 - 9 | 1117 | 40126152 | 74 | |
| BC116835.1 | - | 8 - 9 | 1117 | 40126154 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230309 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230311 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230310 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230313 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230209 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230315 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230210 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101757 | BC116833.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101757 | BC116835.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101757 | BM940984.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101757 | AK144502.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2101757 | BC099382.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 108960 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930361 | AK044446.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930361 | AK054325.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930361 | AK087477.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930361 | BC039952.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930361 | BC106090.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930361 | NM_001010825.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822167 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 339577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 802808 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 894183 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36659 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422205 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 180174 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P113354 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108266 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101757 | BC116833.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101757 | BC116835.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101757 | BM940984.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101757 | AK144502.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2101757 | BC099382.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230323 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230318 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230328 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230320 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230316 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230321 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230305 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230311 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230314 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230325 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230309 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230312 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30230308 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108266 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P113354 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08373544 | - | - |
| Whole Mouse Genome G4122A | A_51_P341041 | BC116833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341041 | BC116835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341041 | BM940984.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341041 | AK144502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P341041 | BC099382.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362603 | BC116833.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362603 | BC116835.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362603 | AK052329.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362603 | AK144502.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P362603 | BC099382.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138096 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105503 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P113354 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108266 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11906797 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11906805 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11906812 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11906809 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11906799 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230209 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230211 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230210 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230213 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230312 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230212 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230309 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230311 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230310 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230314 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230315 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30230313 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230238 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230296 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230229 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230233 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230316 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230186 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230312 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230288 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30230289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230305 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230309 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11883934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230333 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11845559 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230314 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230330 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11948220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230306 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11948218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230335 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230325 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230239 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11906804 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230329 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11852057 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230274 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11873224 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12440947 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12215255 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230299 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230284 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230321 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230238 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11921822 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230318 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12002137 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12283058 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230251 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230301 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230294 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12140120 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230316 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230320 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12427098 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12440952 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11989819 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11906796 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30230296 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10428673 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428679 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428681 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428674 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428675 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428680 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428678 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428676 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10428677 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1452912_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436583_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10428672 | - | - |
| Mouse Genome 430 2.0 Array | 1436583_at | - | - |
| Mouse Genome 430 2.0 Array | 1452912_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1436583_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1452912_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72107_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 72107_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500011625.mm.1 | NM_183089.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500011625.mm.1 | NM_183089.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500011625.mm.1 | TC1500001534.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1436583_PM_at | - | - |
| Mouse Expression Set 430 | 1436583_at | - | - |
| Mouse Expression Set 430 | 1452912_PM_at | - | - |
| Mouse Expression Set 430 | 1452912_at | - | - |
| Murine Genome U74v2 Set | 171219_PM_r_at | - | - |
| Murine Genome U74v2 Set | 165842_PM_at | - | - |
| Murine Genome U74v2 Set | 169188_PM_i_at | - | - |
| Murine Genome U74v2 Set | 171219_r_at | - | - |
| Murine Genome U74v2 Set | 165842_at | - | - |
| Murine Genome U74v2 Set | 169188_i_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436583_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436583_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452912_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1452912_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1770692 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1770692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001770692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1224901 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1224901 | - | - |
| mouse-6 v1.1 (Illumina) | scl47194.8.1_4-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1770692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl47194.8.1_4-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1770692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1224901 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1224901 | - | - |

Dscc1
Genomic Map