Assay Details
| Gene Symbol: Tonsl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183091.3 | NP_898914.3 | 23 - 24 | 3754 | 78 | |
| GenBank mRNA | AK013272.1 | - | 3 - 4 | 552 | 78 | |
| BC066068.1 | - | 23 - 24 | 3736 | 6851399 | 78 | |
More Information
Set Membership: |
|

Tonsl
Genomic Map