Assay Details
| Gene Symbol: Inpp5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127363.1 | NP_001120835.1 | 13 - 14 | 1370 | 69 | |
| NM_183144.3 | NP_898967.2 | 13 - 14 | 1370 | 69 | ||
| GenBank mRNA | AK083449.1 | - | 14 - 15 | 1369 | 69 | |
| AK157558.1 | - | 13 - 14 | 1233 | 69 | ||
| AK171360.1 | - | 13 - 14 | 1370 | 69 | ||
| AK171880.1 | - | 13 - 14 | 1346 | 69 | ||
| BC055471.1 | - | 4 - 5 | 306 | 69 | ||
| BC056341.1 | - | 13 - 14 | 1370 | 5703345 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Inpp5a
Genomic Map