Assay Details
| Gene Symbol: Plk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183152.3 | NP_898975.2 | 3 - 4 | 433 | 61 | |
| GenBank mRNA | AK031968.1 | - | 3 - 4 | 435 | 61 | |
| BB622163.1 | - | 3 - 4 | 434 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK031968.1 transcripts.
Set Membership: |
|

Plk5
Genomic Map