Assay Details
| Gene Symbol: Tmem252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183160.3 | NP_898983.1 | 1 - 2 | 335 | 86 | |
| GenBank mRNA | AK086895.1 | - | 1 - 2 | 337 | 86 | |
| AK142952.1 | - | 1 - 2 | 339 | 86 | ||
| AK143762.1 | - | 1 - 2 | 337 | 86 | ||
| BC087877.1 | - | 1 - 2 | 338 | 30302311 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182026 | AK143762.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182026 | NM_175013.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2182026 | NM_183160.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 433136 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 954357 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777644 | AK038980.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777644 | AK045992.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777644 | XM_137041.6 | refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 777644 | XM_906805.2 | refseq_mrna_mouse_XM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170036 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182026 | AK143762.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182026 | NM_175013.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2182026 | NM_183160.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025030 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025038 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025034 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025027 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025046 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025047 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025024 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025028 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025039 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025032 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025015 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025042 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32025025 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170036 | - | - |
| Whole Mouse Genome G4122A | A_52_P141608 | AK086895.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112503 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170036 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14206514 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14348385 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14152294 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14348388 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14206517 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14348384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025033 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025038 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024987 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025025 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025010 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024978 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025079 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025073 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024997 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025080 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025042 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025044 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025017 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025006 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025077 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32024980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025034 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025059 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025071 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025047 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32025066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32024997 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025063 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14235933 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14444157 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14238372 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025009 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14208370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14155177 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025044 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025067 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025049 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025071 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025014 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32024994 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14175497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025035 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025045 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025034 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025002 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025064 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025066 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14243850 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025046 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025056 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025068 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14208369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025030 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025017 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025084 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025000 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14319919 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025011 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025059 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025086 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14152298 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14206510 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025080 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14206517 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025073 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025025 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025054 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14208371 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32024995 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025033 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025018 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025005 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025076 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32025022 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10462135 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10462134 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10462133 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437595_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10466800 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10462132 | - | - |
| Mouse Genome 430 2.0 Array | 1437595_at | - | - |
| Mouse Genome 430 2.0 Array | 1437595_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 226040_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 226040_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900003645.mm.1 | NM_183160.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003645.mm.1 | NM_183160.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003645.mm.1 | TC1900000397.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900003647.mm.1 | NM_183160.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003647.mm.1 | NM_183160.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003647.mm.1 | TC1900000397.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1900003646.mm.1 | NM_183160.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003646.mm.1 | NM_183160.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900003646.mm.1 | TC1900000397.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437595_at | - | - |
| Mouse Expression Set 430 | 1437595_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437595_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437595_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 828936 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 3850309 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003850309 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2599686 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2599686 | - | - |
| mouse-6 v1.1 (Illumina) | scl53261.2.1_9-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3850309 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl53261.2.1_9-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3850309 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2599686 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2599686 | - | - |

Tmem252
Genomic Map