Assay Details
| Gene Symbol: Fsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183178.2 | NP_899001.1 | 3 - 4 | 367 | 88 | |
| GenBank mRNA | AK140132.1 | - | 3 - 4 | 401 | 88 | |
| BC055106.1 | - | 3 - 4 | 367 | 6532001 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fsd1
Genomic Map