| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200034858.mm.1 |
NM_183180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034858.mm.1 |
NM_183180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034858.mm.1 |
TC0200004154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034868.mm.1 |
NM_183180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034868.mm.1 |
NM_183180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034868.mm.1 |
TC0200004154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034862.mm.1 |
NM_183180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034862.mm.1 |
NM_183180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034862.mm.1 |
TC0200004154.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200034869.mm.1 |
NM_183180.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034869.mm.1 |
NM_183180.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200034869.mm.1 |
TC0200004154.mm.1 |
MTA TC |