Assay Details
| Gene Symbol: Tspan18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183180.2 | NP_899003.1 | 3 - 4 | 382 | 60 | |
| GenBank mRNA | AK032765.1 | - | 2 - 3 | 293 | 60 | |
| BB659903.1 | - | 3 - 4 | 383 | 60 | ||
| BC052169.1 | - | 3 - 4 | 345 | 6335221 | 60 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC052169.1, NM_183180.2 transcripts.
Set Membership: |
|

Tspan18
Genomic Map