Assay Details
| Gene Symbol: Plch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177732.1 | NP_001171203.1 | 11 - 12 | 1592 | 72 | |
| NM_001177733.1 | NP_001171204.1 | 11 - 12 | 1592 | 72 | ||
| NM_183191.3 | NP_899014.2 | 11 - 12 | 1592 | 72 | ||
| GenBank mRNA | AY691172.1 | - | 11 - 12 | 1592 | 72 | |
| AY691173.1 | - | 11 - 12 | 1592 | 72 | ||
| AY691174.1 | - | 11 - 12 | 1592 | 72 | ||
| BC055005.1 | - | 11 - 12 | 1669 | 6491217 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plch1
Genomic Map