| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
NM_001177732.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
NM_001177732.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
NM_001177733.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
NM_001177733.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
NM_183191.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
NM_183191.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015638.mm.1 |
TC0300002071.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
NM_001177732.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
NM_001177732.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
NM_001177733.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
NM_001177733.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
NM_183191.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
NM_183191.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300015641.mm.1 |
TC0300002071.mm.1 |
MTA TC |