Assay Details
| Gene Symbol: Cdkl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183294.2 | NP_899117.1 | 8 - 9 | 968 | 100 | |
| GenBank mRNA | AK016781.1 | - | 8 - 9 | 969 | 100 | |
| BC115909.1 | - | 7 - 8 | 916 | 40105122 | 100 | |
| BC118055.1 | - | 7 - 8 | 917 | 100 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdkl1
Genomic Map