| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_024226.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_024226.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194051.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194051.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194052.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194052.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194053.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194053.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194054.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
NM_194054.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100002560.mm.1 |
TC1100000302.mm.1 |
MTA TC |