Assay Details
| Gene Symbol: Serpinb9d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011460.2 | NP_035590.1 | 6 - 7 | 793 | 128 | |
| GenBank mRNA | AK028382.1 | - | 6 - 7 | 794 | 128 | |
| BC100308.1 | - | 4 - 5 | 601 | 128 | ||
| BC139261.1 | - | 6 - 7 | 769 | 8862283 | 128 | |
| BC139263.1 | - | 6 - 7 | 769 | 8862285 | 128 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971433 | NM_011460.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1971433 | AK028382.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939393 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736203 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616196 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 520220 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 608144 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 299614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 44381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 120829 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160113 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971433 | NM_011460.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1971433 | AK028382.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933264 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933286 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933261 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933276 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933240 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933246 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933268 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933235 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28933283 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160113 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07831361 | - | - |
| Whole Mouse Genome G4122A | A_51_P219424 | BC139261.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P219424 | BC139263.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P219424 | U96704.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P219424 | NM_011460.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P219424 | AK028382.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160113 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10877737 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10620504 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10620511 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10620506 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10620505 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10620517 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933283 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933268 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28933244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10837605 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933261 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933322 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933226 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10620515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933241 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933275 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11030087 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933283 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11102123 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933312 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10837593 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933235 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933311 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11102126 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933279 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933286 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933293 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933232 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10797055 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933270 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P28933276 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P10691855 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10404486 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404483 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404466 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404468 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404457 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404454 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404474 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404481 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404478 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404482 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404455 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404471 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404467 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404473 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404456 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10404475 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457200_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404464 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404452 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10404479 | - | - |
| Mouse Genome 430 2.0 Array | 1457200_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457200_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20726_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20726_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1300002203.mm.1 | NM_011460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002203.mm.1 | NM_011460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002203.mm.1 | TC1300000367.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002200.mm.1 | NM_011460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002200.mm.1 | NM_011460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002200.mm.1 | TC1300000367.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1300002201.mm.1 | NM_011460.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002201.mm.1 | NM_011460.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1300002201.mm.1 | TC1300000367.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457200_PM_at | - | - |
| Mouse Expression Set 430 | 1457200_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457200_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457200_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 600200 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2260136 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2260136 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002260136 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2721641 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2721641 | - | - |
| mouse-6 v1.1 (Illumina) | scl44930.2_243-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2260136 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl44930.2_243-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2260136 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2919035 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2919034 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2919034 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2919035 | - | - |

Serpinb9d
Genomic Map