Assay Details
| Gene Symbol: Gpr151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181543.1 | NP_853521.1 | 1 | 1589 | 148 | |
| GenBank mRNA | AJ564167.1 | - | 1 | 1589 | 148 | |
| AK048591.1 | - | 1 | 1247 | 148 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914335 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P100263 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656229 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656221 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656263 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656209 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656211 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656244 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656226 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656220 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656212 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656223 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656230 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656214 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656228 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656232 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656239 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656252 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656250 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656236 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656218 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656222 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656216 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656254 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656231 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656258 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656249 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656234 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656242 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31656262 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P100263 | - | - |
| Whole Mouse Genome G4122A | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | BC120520.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | BC120526.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | NM_181543.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | AJ564167.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P240703 | AK048591.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127834 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P100263 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13793220 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14016701 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14016715 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14016727 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13793222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656276 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656228 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31656211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656214 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13999896 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656264 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656244 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14016734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656263 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656242 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656252 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656236 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656211 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656228 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656207 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656220 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656209 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14021039 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656281 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656280 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656234 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656231 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14016707 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656218 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13973163 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13967766 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656240 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656268 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656208 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14016725 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656277 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656216 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656230 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656212 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31656232 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10458642 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458644 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10458643 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1457555_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10458641 | - | - |
| Mouse Genome 430 2.0 Array | 1457555_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1457555_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240239_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 240239_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1800009336.mm.1 | NM_181543.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009336.mm.1 | NM_181543.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1800009336.mm.1 | TC1800001259.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1457555_at | - | - |
| Mouse Expression Set 430 | 1457555_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457555_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1457555_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 298038 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2850280 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2850280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002850280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1242331 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1242331 | - | - |
| mouse-6 v1.1 (Illumina) | scl51467.1.342_127-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2850280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl51467.1.342_127-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2850280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1242331 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1242331 | - | - |

Gpr151
Genomic Map