Assay Details
| Gene Symbol: Lypd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182785.3 | NP_877586.1 | 3 - 4 | 490 | 61 | |
| GenBank mRNA | AK077071.1 | - | 3 - 4 | 491 | 61 | |
| BC049744.1 | - | 2 - 3 | 290 | 6772266 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25006992 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25006991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25006990 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393738 | NM_182785.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393738 | AK077071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393738 | BC049744.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393738 | NM_182785.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393738 | AK077071.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P393738 | BC049744.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893017 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 760503 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891584 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169923 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157718 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393738 | NM_182785.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393738 | AK077071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393738 | BC049744.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393738 | NM_182785.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393738 | AK077071.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P393738 | BC049744.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006975 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006980 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006978 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006987 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006983 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006958 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006976 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006974 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006982 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25006961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157718 | - | - |
| Whole Mouse Genome G4122A | A_52_P393738 | NM_182785.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393738 | AK077071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393738 | BC049744.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393738 | NM_182785.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393738 | AK077071.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P393738 | BC049744.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112785 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157718 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06625979 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06625992 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206965 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206967 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06625988 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06445603 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05935916 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06244974 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06445600 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06445605 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06625976 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206968 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206970 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06445606 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06206969 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25006991 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25006990 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25006992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006961 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006932 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006954 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006921 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006963 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006919 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006976 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006938 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006918 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006937 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006940 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25007039 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006915 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006992 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006966 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006912 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006943 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006924 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25006987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445600 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06201261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006952 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06201266 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206965 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006962 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006991 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06445601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006961 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006960 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06244974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006978 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05935916 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06201260 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06206968 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006943 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006953 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006956 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006940 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06201265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06625992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006950 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006987 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06625986 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25006941 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10560906 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560905 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560904 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10560907 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437596_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10560903 | - | - |
| Mouse Genome 430 2.0 Array | 1437596_at | - | - |
| Mouse Genome 430 2.0 Array | 1437596_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232973_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 232973_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700022530.mm.1 | TC0700002612.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700022530.mm.1 | NM_182785.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700022530.mm.1 | NM_182785.3 | Refseq |
| Mouse Expression Set 430 | 1437596_PM_at | - | - |
| Mouse Expression Set 430 | 1437596_at | - | - |
| Murine Genome U74v2 Set | 165595_PM_at | - | - |
| Murine Genome U74v2 Set | 165595_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437596_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437596_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 700213 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6180400 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180400 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2600135 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2600135 | - | - |
| mouse-6 v1.1 (Illumina) | scl31645.4.1_60-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180400 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31645.4.1_60-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180400 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2600135 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2600135 | - | - |

Lypd4
Genomic Map