Assay Details
| Gene Symbol: Bivm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144558.4 | NP_653141.3 | 3 - 4 | 1028 | 106 | |
| GenBank mRNA | AF411386.1 | - | 4 - 5 | 1031 | 106 | |
| AK032375.1 | - | 3 - 4 | 988 | 106 | ||
| AK034329.1 | - | 3 - 4 | 990 | 106 | ||
| BC082584.1 | - | 3 - 4 | 834 | 30612339 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bivm
Genomic Map