Assay Details
| Gene Symbol: Zfp184 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183014.1 | NP_898835.1 | 3 - 4 | 416 | 60 | |
| GenBank mRNA | AK036164.1 | - | 3 - 4 | 401 | 60 | |
| BC053084.1 | - | 3 - 4 | 416 | 5708591 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zfp184
Genomic Map