Assay Details
| Gene Symbol: Ttll12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183017.2 | NP_898838.2 | 3 - 4 | 581 | 88 | |
| GenBank mRNA | AK084879.1 | - | 3 - 4 | 546 | 89 | |
| AK134345.1 | - | 3 - 4 | 547 | 88 | ||
| AK143496.1 | - | 2 - 3 | 412 | 88 | ||
| AK150116.1 | - | 3 - 4 | 547 | 88 | ||
| AM690756.1 | - | 3 - 4 | 534 | 88 | ||
| BC055368.1 | - | 3 - 4 | 546 | 30055801 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll12
Genomic Map