Assay Details
| Gene Symbol: Ttll12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183017.2 | NP_898838.2 | 6 - 7 | 943 | 98 | |
| GenBank mRNA | AK084879.1 | - | 6 - 7 | 99 | ||
| AK134345.1 | - | 6 - 7 | 910 | 98 | ||
| AK143496.1 | - | 5 - 6 | 775 | 98 | ||
| AK150116.1 | - | 6 - 7 | 909 | 98 | ||
| AM690756.1 | - | 6 - 7 | 896 | 98 | ||
| BC055368.1 | - | 6 - 7 | 908 | 30055801 | 98 | |
More Information
Set Membership: |
|

Ttll12
Genomic Map