Assay Details
| Gene Symbol: Ttll12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183017.2 | NP_898838.2 | 7 - 8 | 1063 | 67 | |
| GenBank mRNA | AK084879.1 | - | 7 - 8 | 1030 | 67 | |
| AK134345.1 | - | 7 - 8 | 1030 | 67 | ||
| AK143496.1 | - | 6 - 7 | 895 | 67 | ||
| AK150116.1 | - | 7 - 8 | 1029 | 67 | ||
| AM690756.1 | - | 7 - 8 | 1016 | 67 | ||
| BC055368.1 | - | 7 - 8 | 1028 | 30055801 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttll12
Genomic Map