Assay Details
| Gene Symbol: Trmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164559.1 | NP_001158031.1 | 12 - 13 | 1518 | 58 | |
| NM_001164560.1 | NP_001158032.1 | 12 - 13 | 1442 | 58 | ||
| NM_198020.2 | NP_932137.2 | 12 - 13 | 1545 | 58 | ||
| GenBank mRNA | AK032945.1 | - | 10 - 11 | 2978 | 58 | |
| AK084775.1 | - | 12 - 13 | 1485 | 58 | ||
| AK155251.1 | - | 12 - 13 | 1511 | 58 | ||
| AK159467.1 | - | 12 - 13 | 1483 | 58 | ||
| AK170412.1 | - | 12 - 13 | 1407 | 58 | ||
| AK171002.1 | - | 12 - 13 | 1510 | 58 | ||
| AK212736.1 | - | 1 - 2 | 81 | 58 | ||
| BC027404.1 | - | 12 - 13 | 1514 | 4950980 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt1
Genomic Map