Assay Details
| Gene Symbol: Trmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164559.1 | NP_001158031.1 | 15 - 16 | 1822 | 78 | |
| NM_001164560.1 | NP_001158032.1 | 15 - 16 | 1746 | 78 | ||
| NM_198020.2 | NP_932137.2 | 15 - 16 | 1849 | 78 | ||
| GenBank mRNA | AK032945.1 | - | 13 - 14 | 3282 | 78 | |
| AK155251.1 | - | 15 - 16 | 1815 | 78 | ||
| AK159467.1 | - | 15 - 16 | 1787 | 78 | ||
| AK170412.1 | - | 15 - 16 | 1711 | 78 | ||
| AK171002.1 | - | 15 - 16 | 1814 | 78 | ||
| BB055133.2 | - | 3 - 4 | 377 | 78 | ||
| BC027404.1 | - | 15 - 16 | 1818 | 4950980 | 78 | |
More Information
Set Membership: |
|

Trmt1
Genomic Map