Assay Details
| Gene Symbol: Pnlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026925.3 | NP_081201.2 | 11 - 12 | 1200 | 109 | |
| GenBank mRNA | AY387690.1 | - | 10 - 11 | 1172 | 109 | |
| BC061061.1 | - | 11 - 12 | 1200 | 30316303 | 109 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pnlip
Genomic Map