Assay Details
| Gene Symbol: Ssh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177710.4 | NP_808378.2 | 5 - 6 | 430 | 85 | |
| GenBank mRNA | AB099288.1 | - | 5 - 6 | 430 | 85 | |
| AK170142.1 | - | 4 - 5 | 563 | 85 | ||
| BC141392.1 | - | 5 - 6 | 424 | 9056005 | 85 | |
| BC141393.1 | - | 5 - 6 | 424 | 9056006 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ssh2
Genomic Map