Assay Details
| Gene Symbol: Gnl3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168600.1 | NP_001162071.1 | 14 - 15 | 1562 | 100 | |
| NM_198110.2 | NP_932778.1 | 14 - 15 | 1595 | 100 | ||
| GenBank mRNA | BC020354.1 | - | 14 - 15 | 1531 | 100 | |
| BC057033.1 | - | 14 - 15 | 1521 | 6834283 | 100 | |
| BC079653.1 | - | 14 - 15 | 1517 | 6843880 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gnl3l
Genomic Map