Assay Details
| Gene Symbol: Lrrc42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029985.2 | NP_084261.1 | 5 - 6 | 1090 | 102 | |
| GenBank mRNA | AK020847.1 | - | 5 - 6 | 896 | 102 | |
| AK169814.1 | - | 3 - 4 | 548 | 102 | ||
| BC027203.1 | - | 5 - 6 | 1022 | 3710091 | 102 | |
| BC031894.1 | - | 5 - 6 | 994 | 5362699 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc42
Genomic Map