Assay Details
| Gene Symbol: Mtmr9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177594.1 | NP_808262.1 | 4 - 5 | 904 | 63 | |
| GenBank mRNA | AK035026.1 | - | 4 - 5 | 919 | 63 | |
| AK044957.1 | - | 4 - 5 | 934 | 63 | ||
| AK152320.1 | - | 4 - 5 | 690 | 63 | ||
| BC046275.1 | - | 4 - 5 | 904 | 6307469 | 63 | |
| BC145946.1 | - | 4 - 5 | 659 | 40131249 | 63 | |
| BC145948.1 | - | 4 - 5 | 659 | 40131251 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mtmr9
Genomic Map