Assay Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285991.1 | NP_001272920.1 | 13 - 14 | 1340 | 79 | |
| NM_001285993.1 | NP_001272922.1 | 13 - 14 | 1307 | 79 | ||
| NM_028162.4 | NP_082438.3 | 13 - 14 | 1340 | 79 | ||
| GenBank mRNA | AK033710.1 | - | 13 - 14 | 1232 | 79 | |
| AK144467.1 | - | 13 - 14 | 1341 | 79 | ||
| AK154975.1 | - | 13 - 14 | 1300 | 79 | ||
| AK220251.1 | - | 13 - 14 | 1307 | 79 | ||
| BC043113.1 | - | 13 - 14 | 1156 | 6407234 | 79 | |
More Information
Set Membership: |
|

Tbc1d5
Genomic Map