Assay Details
| Gene Symbol: Tbc1d5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285991.1 | NP_001272920.1 | 15 - 16 | 1600 | 98 | |
| NM_001285993.1 | NP_001272922.1 | 15 - 16 | 1567 | 98 | ||
| NM_028162.4 | NP_082438.3 | 15 - 16 | 1600 | 98 | ||
| GenBank mRNA | AK144467.1 | - | 15 - 16 | 1601 | 98 | |
| AK154975.1 | - | 15 - 16 | 1560 | 98 | ||
| AK163366.1 | - | 15 - 16 | 1629 | 98 | ||
| AK220251.1 | - | 15 - 16 | 1567 | 98 | ||
| BC043113.1 | - | 15 - 16 | 1416 | 6407234 | 98 | |
| BC098328.1 | - | 10 - 11 | 946 | 30357143 | 98 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tbc1d5
Genomic Map